Learn More
Motic™ LED Ring Light
Product Code. 11808753
Change view
Click to view available options
For Use With (Equipment):
SMZ Series Miroscopes without incident light
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11808753 SMZ Series Miroscopes without incident light Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11808753 Supplier Motic™ Supplier No. 1101002402721
  • ONLINE EXCLUSIVE PRICE
    196.00 EUR
    175.85 EUR / Pack of 1
    Save 20.15 EUR
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Screws on to SMZ series stereo microscopes. Motic™ LED Ring Light is compatible only with SMZ models supplied without incident light
  • 60T
  • Dimmable and segmentable
  • For use only with SMZ Stereo Microscope without incident light
Specifications
Type LED Ring Illumination 60T
Description dimmable, segmentable
For Use With (Equipment) SMZ Series Miroscopes without incident light
Quantity 1 Ea.