Learn More
Motic™ Spare Bulb for Microscope
Product Code. 15398292
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. 15398292
Brand: Motic™ 1101002600161
Only null left
Add to basket
Add to basket
Replacement Bulb: LED, 20mA, 3.5V, 70mW
  • Replacement bulb for microscope
  • LED
  • 20mA
  • 3.5V
  • 70mW
Order Info
dimProductType
Specifications
Type Replacement bulb
Quantity 1 Ea.
Voltage 3.5 V