Learn More
Berthold Technologies LED Plant Growth Illumination Far-red/R/G/B
Product Code. 31777393 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
LED Panel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31777393 LED Panel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31777393 Supplier Berthold Technologies Supplier No. 5658901
 Restricted Item
Estimated Shipment: 20-11-2026
LED illumination for NightSHADE imaging systems that enables daylight simulation with customizable settings.
  • Two LED panels, each with four colors
  • Individual control of LED intensity and duration
  • Supports simulation of daylight with spectral and intensity gradients
  • Equipped with the following colors: Far red, Red, Green, Blue
Specifications
Type LED Panel
For Use With (Application) Daylight Simulation
For Use With (Equipment) LED Panel
Voltage 24 V