Learn More
Invitrogen™ Lecanemab Recombinant Monoclonal Antibody
Product Code. 30283188
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30283188 100 μg 100µg
30283577 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30283188 Supplier Invitrogen™ Supplier No. MA559917
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lecanemab, marketed as Leqembi, is a medication that is prescribed to treat Alzheimer's disease. Lecanemab is a monoclonal antibody derived from a mouse antibody called mAb158, which has been humanized to improve its compatibility with humans. It specifically targets amyloid beta, a protein associated with the disease.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Lecanemab Humanized
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Gene Alias BAN2401; Leqembi
Host Species Human
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ