Learn More
Fisherbrand™ Leaf Calibration Weight Set, Class E2 with certificate
Product Code. 15515119 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
1mg to 500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15515119 1mg to 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15515119
  • ONLINE EXCLUSIVE PRICE
    793.00 EUR
    640.00 EUR / 1 units
    Save 153.00 EUR
    19% Off
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
  • Delivered with a calibration certificate guaranteeing traceability
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Classification E2
Weight (Metric) 1 to 500 mg
Mass 1 mg to 500 mg