Learn More
Lead(II) Oxide, For Analysis, EMSURE™, MilliporeSigma™
Product Code. 15854980
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15854980 1 kg Plastic bottle 1kg
15844980 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15854980 Supplier Supelco Supplier No. 1074011000
 Restricted Item
Estimated Shipment: 26-11-2026
For analysis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name plumbanone
SMILES O=[Pb]
Specifications
Physical Form Solid
Quantity 1 kg
Odor Odorless
Packaging Plastic bottle
Appearance Yellow solid
UN Number UN 2291
Formula Weight 223.19
Percent Purity ≥99.0%
Chemical Name or Material Lead(II) oxide