missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) nitrate, MilliporeSigma™
Specifications
Specifications
| Boiling Point | >500°C |
| CAS | 10099-74-8 |
| Quantity | 100 g |
| Molecular Formula | N2O6Pb |
| MDL Number | MFCD00011153 |
| UN Number | UN1469 |
| Synonym | lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt |
| Solubility Information | Soluble in water: 522g/L at 20°C |
| InChI Key | RLJMLMKIBZAXJO-UHFFFAOYSA-N |
| SMILES | [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?