missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Carbonate, Supelco™

Product Code. 15807580
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15807580 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
Product Code. 15807580 Supplier Supelco Supplier No. 1073810250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement

For analysis EMSURE™ ACS

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 315°C (Decomposition)
CAS 598-63-0
Quantity 250 g
MDL Number MFCD00064631
Synonym Lead(II) carbonate, Lead(II) carbonate
Molecular Weight (g/mol) 267.21
Grade Research
Density 6.6 g/cm3 at 20°C
Chemical Name or Material Lead(II) Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.