missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Acetate Trihydrate, MilliporeSigma™, For Analysis, EMSURE™, MilliporeSigma™

Product Code. 15824980
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15824980 250 g Plastic Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15824980 Supplier Supelco Supplier No. 1073750250
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 75°C
CAS 6080-56-4
pH 5.5 to 6.5 (50g/L, H2O, 20°C)
Quantity 250 g
Molecular Formula C4H12O7Pb
MDL Number MFCD00150023
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
Solubility Information 443g/L
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
IUPAC Name λ²-lead(2+) diacetate trihydrate
Molecular Weight (g/mol) 379.30
PubChem CID 22456
ChEBI CHEBI:33112
Formula Weight 379.34
Percent Purity 99.5 to 102.0%
Packaging Plastic Bottle
Density 3.3 g/3 (20 °C)
Chemical Name or Material Lead(II) Acetate Trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.