Learn More
Lead(II) 2,4-pentanedionate
Product Code. 11423840
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423840 10 g 10g
11433840 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11423840 Supplier Thermo Scientific Alfa Aesar Supplier No. 089858.09
 Restricted Item
In Stock
Add to basket
CAS: 15282-88-9 | C10H14O4Pb | 405.418 g/mol
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15282-88-9
Molecular Formula C10H14O4Pb
Molecular Weight (g/mol) 405.418
MDL Number MFCD00013499
InChI Key UNNUWSQNTAFLDC-SYWGCQIGSA-L
Synonym lead ii acetylacetonate, 2,4-pentanedione lead ii derivative
PubChem CID 53393517
IUPAC Name bis[[(E)-4-oxopent-2-en-2-yl]oxy]lead
SMILES CC(=CC(=O)C)O[Pb]OC(=CC(=O)C)C
Specifications
Melting Point 141°C to 144°C
Odor Odorless
Quantity 10 g
UN Number UN3467
Sensitivity Hygroscopic
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 405.41
Physical Form Crystalline
Chemical Name or Material Lead (II) 2,4-pentanedionate
compliance-icons compliance-icons
Product Identifier
  • Lead(II) 2,4-pentanedionate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..