missing translation for 'onlineSavingsMsg'
Learn More

Hach™ LCK Cuvette Tests

Product Code. 10749873
Change view
Click to view available options
Analytes:
Nitrite
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Analytes Unit Size
10749873 Nitrite Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10749873 Supplier Hach™ Supplier No. LCK541
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Insert, read and finish. Hach™ LCK Cuvette Tests are pre-dosed reagents packaged in a fully equipped box. They provide maximum safety, ease of use and photometric evaluation, while using a minimum amount of reagent/chemicals for an environmentally friendly solution.

Hach LCK Cuvette Tests are officially approved for legally required consent limits. With the help of Addista standard solutions and round robin test solutions, they provide the assurance that analytical results are always correct. The photometer identifies the cuvette test from its barcode (IBR+) and calculates the result immediately. Cuvettes provide precision and safety using the Dosicap Zip that eliminated contact with reagents.

  • Pre-dosed, ready-to-use reagents with no rinsing steps needed
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding

Order Info

dimRange

TRUSTED_SUSTAINABILITY

Specifications

Analytes Nitrite
Certifications/Compliance EN ISO 26777, DIN 38405 D10
Color Green
Format Pre-dosed Tubes
For Use With (Application) Water quality testing
For Use With (Equipment) Photometer (DR3900, DR6000, DR2800, DR3800, DR5000), Addista standard solutions
Includes Fully equipped box, reagent set and LZP341 (50mm semi-micro plastic cuvettes, 10 pcs)
No. of Tests 50
Range 0.0015 to 0.03mg/L NO2-N
Storage Requirements 2°C to 8°C
Test Method Diazotization
Type Nitrite Trace Cuvette Test
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.