Learn More
Testo™ LCD Display Thermo-Hygrometer
Product Code. 10316802
Change view
Click to view available options
Temperature Range (Metric):
0°C to 50°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
10316802 0°C to 50°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10316802 Supplier Testo™ Supplier No. 0560 6081
Only null left
Add to basket
Add to basket
Use this thermohygrometer for a variety of applications. Testo™ 608-H1 Thermohygrometers provide a continuous display of current temperature and humidity values.
  • Continuous measurement and display of temperature and relative humidity
  • Calculation and display of the dew point
  • Large, clear display
  • Capacitive humidity sensor with long-term stability for many years of measurement
Specifications
Type Thermohygrometer
Resolution 0.001
Temperature Range (Metric) 0°C to 50°C
Display Type LCD
Weight (Metric) 168 g
Voltage 9 V
For Use With (Application) For measure and display the temperature and relative humidity continuously