missing translation for 'onlineSavingsMsg'
Learn More

Lazertinib, MedChemExpress

Product Code. 16470597
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470597 50 mg 50mg
16466987 5 mg 5mg
16465196 100 mg 100mg
16454268 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470597 Supplier Medchem Express Supplier No. HY109061

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lazertinib (YH25448) is a potent, highly mutant-selective, blood-brain barrier permeable, orally available and irreversible third-generation EGFR tyrosine kinase inhibitor, and can be used in the research of non-small cell lung cancer.

Lazertinib (YH25448) is a potent, highly mutant-selective, blood-brain barrier permeable, orally available and irreversible third-generation EGFR tyrosine kinase inhibitor, and can be used in the research of non-small cell lung cancer.

Specifications

Chemical Name or Material Lazertinib
CAS 1903008-80-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H34N8O3
Quantity 50 mg
Synonym YH25448 GNS-1480
Solubility Information DMSO : 9 mg/mL (16.23 mM; Need ultrasonic)
SMILES C=CC(NC1=CC(NC2=NC=CC(N3N=C(C4=CC=CC=C4)C(CN(C)C)=C3)=N2)=C(OC)C=C1N5CCOCC5)=O
Molecular Weight (g/mol) 554.64
Formula Weight 554.64
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.