Learn More
Lazertinib, MedChemExpress
Product Code. 16466987
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466987 5 mg 5mg
16470597 50 mg 50mg
16465196 100 mg 100mg
16454268 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16466987 Supplier Medchem Express Supplier No. HY109061
MedChemExpress Lazertinib (YH25448) is a potent, highly mutant-selective, blood-brain barrier permeable, orally available and irreversible third-generation EGFR tyrosine kinase inhibitor, and can be used in the research of non-small cell lung cancer.
Lazertinib (YH25448) is a potent, highly mutant-selective, blood-brain barrier permeable, orally available and irreversible third-generation EGFR tyrosine kinase inhibitor, and can be used in the research of non-small cell lung cancer.
Specifications
Chemical Name or Material Lazertinib
CAS 1903008-80-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H34N8O3
Quantity 5 mg
Synonym YH25448 GNS-1480
Solubility Information DMSO : 9 mg/mL (16.23 mM; Need ultrasonic)
SMILES C=CC(NC1=CC(NC2=NC=CC(N3N=C(C4=CC=CC=C4)C(CN(C)C)=C3)=N2)=C(OC)C=C1N5CCOCC5)=O
Molecular Weight (g/mol) 554.64
Formula Weight 554.64
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease