Learn More
Lauroylcholine Chloride, TRC
Product Code. 30471457
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471457 5 g 5g
30481803 1 g 1g
30472147 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30471457 Supplier Toronto Research Chemicals Supplier No. TRCL1850005G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25234-60-0
Molecular Formula C17 H36 N O2 . Cl
Molecular Weight (g/mol) 321.93
IUPAC Name 2-dodecanoyloxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCC(=O)OCC[N+](C)(C)C
Specifications
Quantity 5 g
InChI Formula InChI=1S/C17H36NO2.ClH/c1-5-6-7-8-9-10-11-12-13-14-17(19)20-16-15-18(2,3)4;/h5-16H2,1-4H3;1H/q+1;/p-1
Formula Weight 321.2435
Chemical Name or Material Lauroylcholine Chloride