missing translation for 'onlineSavingsMsg'
Learn More

Lauric Acid Methyl Ester, Technical Grade, 99.1%, MP Biomedicals™

Product Code. 11401882 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401882 100 g 100g
11491872 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11401882 Supplier MP Biomedicals Supplier No. 0210526380
  • ONLINE EXCLUSIVE PRICE
    578.00 EUR
    550.20 EUR / 100g
    Save 27.80 EUR
    4% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Insoluble in water

Chemical Identifiers

CAS 111-82-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
InChI Key UQDUPQYQJKYHQI-UHFFFAOYSA-N
Synonym methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
PubChem CID 8139
ChEBI CHEBI:87494
IUPAC Name methyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC

Specifications

Color Colorless
Assay Percent Range 99.1%
Quantity 100 g
Formula Weight 214.3
Grade Technical
Physical Form Liquid
Chemical Name or Material Lauric Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.