Learn More
Fisherbrand™ Latex Dropper Teats
Product Code. 12426180 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
1 mL
1.5 mL
1.5 to 5 mL (Assorted)
2.5 mL
3.75 mL
Unit Size:
Pack of 20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12426180 2.5 mL Pack of 20
12406180 1 mL Pack of 20
12416180 1.5 mL Pack of 20
12436180 3.75 mL Pack of 20
12446180 1.5 to 5 mL (Assorted) Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12426180
  • ONLINE EXCLUSIVE PRICE
    27.33 EUR
    12.56 EUR / Pack of 20
    Save 14.77 EUR
    54% Off
View all promotions
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
For easy dispensing of liquids.
  • Flexible rubber bulbs create suction and dispensing capacity when used with pipettes
  • Provide intuitive manual pipeting control
  • Available in different capacities
Specifications
Material Latex
Type Bulb Dropper
Volume (Metric) 2.5 mL
Diameter (Metric) Opening 6 mm