Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Clostridium difficile
Shop All Novacyt Group ProductsBeskrivelse
- From a range of available technologies for common pathogens and causatives of bowel disease.
- Specific tests available for a variety of infection detections.
- Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
- Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
- Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Tekniske data
Tekniske data
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | C. difficile |
| Type | Latex Agglutination Test |
| Quantity | 50 Tests |
| Test Time | 2 min. |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?