Learn More
Invitrogen™ LanthaScreen™ Tb-labeled Anti-Mouse Ab
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
Description
- A general reagent for use in assay development
- This terbium labeled antibody will bind mouse antibodies
- It is ideally suited for use during assay development when evaluating multiple primary mouse antibodies
- Also ideal for use when it is impossible to directly terbium label a primary antibody
Specifications
Specifications
| Content And Storage | Terbium-labeled anti-Mouse antibody in HEPES buffered saline. Store at -20°C. |
| Excitation/Emission | 340/490, 520 |
| Quantity | 1 mg |
| Reactivity | Mouse |
| Readout | End Point |
| Shipping Condition | Dry Ice |
| Conjugate | Lanthanide Chelates, Terbium (Tb) |
| Product Line | LanthaScreen |