missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) Chloride Heptahydrate 98%, Supelco™
Specifications
Specifications
| Melting Point | 91°C (Elimination of Water of Crystallisation) |
| CAS | 10025-84-0 |
| Quantity | 250 g |
| MDL Number | MFCD00149756 |
| Synonym | Lanthanum(III) chloride heptahydrate 98%, Lanthanum trichloride, Lanthanum(III) chloride heptahydrate 98% |
| Molecular Weight (g/mol) | 371.37 |
| Grade | Research |
| Chemical Name or Material | Lanthanum(III) Chloride Heptahydrate 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?