missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride heptahydrate 98%, MilliporeSigma™

Product Code. 15885630
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15885630 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15885630 Supplier Supelco Supplier No. 1122190100
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity grade lanthanum(III) chloride heptahydrate for analytical applications.

  • Lanthanum (III) chloride heptahydrate 98%, reagent for analysis, supplied in a 100 g plastic bottle
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 91°C
CAS 10025-84-0, 7732-18-5
Color White
pH 5
Physical Form Solid
Quantity 100 g
Assay Percent Range 98%
Molecular Formula Cl3H14LaO7
Linear Formula LaCl3 · 7H2O
MDL Number MFCD00149756
Synonym Lanthanum trichloride
Solubility Information Soluble (20°C)
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
IUPAC Name lanthanum(3+) heptahydrate trichloride
Molecular Weight (g/mol) 371.36
Formula Weight 371.37
Identification Passes Test
Packaging Plastic Bottle
Appearance White Solid
Density Bulk Density: 900 kg/m3
Chemical Name or Material For Analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.