Learn More
3M™ Lane and Safety Marking Tape 471
Product Code. 17701699
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Unit Size:
12 rolls
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17701699 White 12 rolls
17781679 Red 12 rolls
17741689 Yellow 12 rolls
17711689 Green 12 rolls
17791679 Blue 12 rolls
17701689 Black 12 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17701699 Supplier 3M™ Supplier No. 7100260598
  • ONLINE EXCLUSIVE PRICE
    846.00 EUR
    704.07 EUR / 12 rolls
    Save 141.93 EUR
    16% Off
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
Strength, conformability, instant adhesion and long-term clean removal make this tape an ideal choice for all but the heaviest floor and safety marking applications. Commonly used for marking hazards and aisles, color-coding, paint masking, decorative trim, and splicing, this tape comes individually wrapped and conveniently packaged.
  • Maintains color when exposed to abrasion, wear and many solvents
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, high holding strength on many surfaces
  • Available in many colors (yellow, white, red, black, brown, green, orange, purple, blue and transparent)
  • Clean removal from many surfaces to help reduce clean-up and labor costs
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 33 m x 50 mm
Packaging Quantity 12 Rolls
Thickness 0.14 mm
Adhesive Type Rubber-based Adhesive
Color White
Material Vinyl