Learn More
Lamotrigine N-Acetate, TRC
Product Code. 30468526
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468526 250 mg 250mg
30477864 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30468526 Supplier Toronto Research Chemicals Supplier No. TRCL173255250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 77668-57-6
Molecular Formula C11 H9 Cl2 N5 O
Molecular Weight (g/mol) 298.13
Synonym Acetamide, N-[3-amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5-yl]-, N5-Acetyllamotrigine
IUPAC Name N-[3-amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5-yl]acetamide
SMILES CC(=O)Nc1nc(N)nnc1c2cccc(Cl)c2Cl
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C11H9Cl2N5O/c1-5(19)15-10-9(17-18-11(14)16-10)6-3-2-4-7(12)8(6)13/h2-4H,1H3,(H3,14,15,16,18,19)
Formula Weight 297.0184
Chemical Name or Material Lamotrigine N-Acetate