Learn More
Brady™ Laminated Polyester Label Markers
Product Code. 11795563 Shop All Brady Products
Change view
Click to view available options
Color:
Black on Gray
White on Green
White on Purple
White on Red
Height (Metric) Character:
8 mm
13 mm
Unit Size:
1 roll
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Height (Metric) Character Unit Size
11795563 White on Purple 13 mm 1 roll
11785563 Black on Gray 13 mm 1 roll
11765563 White on Green 13 mm 1 roll
11745583 Black on Gray 8 mm 1 roll
11725583 White on Green 8 mm 1 roll
11755583 White on Purple 8 mm 1 roll
11745563 White on Red 13 mm 1 roll
11705583 White on Red 8 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11795563 Supplier Brady™ Supplier No. 273514
  • ONLINE EXCLUSIVE PRICE
    349.00 EUR
    325.09 EUR / 1 rolls
    Save 23.91 EUR
    6% Off
Estimated Shipment: 13-11-2026
Laminated Polyester Label Markers feature a surface printed white polyester film with a permanent, acrylic based, pressure sensitive adhesive and over-laminated with a clear polyester film.
  • Used for pipe markers and safety signs.
  • B-7541 gives excellent adhesion to low surface energy surfaces, such as polypropylene and ABS, as well as on most powder coatings.
  • Standard ink colors are black, red (RAL 3001-GL), yellow (RAL 1003-GL), blue (RAL5005-GL) and green (RAL 6032-GL).
Specifications
Adhesive Type Permanent Acrylic
Color White on Purple
Finish Gloss
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Acide
For Use With (Application) Pipe and valve marking
Languages French
No. per Pack 220 Markers/Roll
Quantity 220 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 33 m x 100 mm
Dimensions (L x W x H) 33 m x 100 mm x 150 mm
Length (English) 108.27 ft.
Length (Metric) 33 m
Width (English) 3.94 in.
Width (Metric) 100 mm
Height (English) 5.91 in.
Height (Metric) 150 mm
Height (Metric) Character 13 mm
Diameter (English) 3 in.
Diameter (Metric) 70 mm
Alternate Part Number PMF-ACIDE-PR4-RLL090