Learn More
Brady™ Laminated Polyester ADR 1.5 1 Explosive Substances Transport Signs
Product Code. 11740204
Change view
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Width (Metric):
100.00 mm
200.00 mm
297.00 mm
Quantity:
1 Ea.
250/Pk.
Unit Size:
1 roll
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Quantity unitSize
11740204 100 mm 100.00 mm 1 Ea. Each
11790234 100 mm 100.00 mm 250/Pk. 1 roll
11750204 200 mm 200.00 mm 1 Ea. Each
11760204 297 mm 297.00 mm 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11740204 Supplier Brady™ Supplier No. 223555
  • ONLINE EXCLUSIVE PRICE
    4.10 EUR
    3.77 EUR / Pack of 1
    Save 0.33 EUR
    8% Off
Estimated Shipment: 20-08-2026
Specifications
Alternate Part Number ADR 1.5-100*100-B7541
Finish Gloss
Material Laminated Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100.00 mm
Temperature Range (Metric) -40 °C-110 °C
Shape Diamond
Displayed Text 1.5 1
Color Black on Orange