Learn More
Uvex™ Ladies Coat 7463-White
Well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove.
78.30 EUR
Specifications
| Gender | Women's |
|---|---|
| Material | Cotton |
| Color | White |
| Closure Type | Press Studs |
| Disposable | Reusable |
| Product Code | Brand | Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Size | Price | Quantity & Availability | |||||
|
17215701
|
Uvex™
8825808 |
X-Small |
78.30 EUR
Pack of 1 |
Estimated Shipment: 09-06-2026
Log in to see stock. |
|||||
|
17225701
|
Uvex™
8825809 |
Small |
78.30 EUR
Pack of 1 |
Estimated Shipment: 09-06-2026
Log in to see stock. |
|||||
|
17235701
|
Uvex™
8825810 |
Medium |
78.30 EUR
Pack of 1 |
Estimated Shipment: 04-06-2026
Log in to see stock. |
|||||
|
17245701
|
Uvex™
8825811 |
Large |
78.30 EUR
Pack of 1 |
Estimated Shipment: 09-06-2026
Log in to see stock. |
|||||
|
17255701
|
Uvex™
8825812 |
X-Large |
78.30 EUR
Pack of 1 |
Estimated Shipment: 09-06-2026
Log in to see stock. |
|||||
|
17265701
|
Uvex™
8825813 |
2X-Large |
78.30 EUR
Pack of 1 |
Estimated Shipment: 09-06-2026
Log in to see stock. |
|||||
|
17275701
|
Uvex™
8825814 |
3X-Large |
78.30 EUR
Pack of 1 |
Estimated Shipment: 09-06-2026
Log in to see stock. |
|||||
|
17285701
|
Uvex™
8825815 |
4X-Large |
78.30 EUR
Pack of 1 |
Estimated Shipment: 09-06-2026
Log in to see stock. |
|||||
Description
- Women coat
- Material: 66.5% polyester, 32% cotton, 1.5% antistatic. Fibers, 245 g / m²
- Color white
- Highest protection with antistat.
- Fiber - optimal charge reduction
- High rise arm construction
- Breast pocket on the left
- Two side pockets
- Turn-down collar standards: IEC 61340, OEKO-TEX™ Standard 100
- Sizes: XS-4XL
- Normal wash 60 ° C, do not bleach, dry Dryer, iron 2 points, chem. Cleaning mi perchlorethylene
Specifications
| Women's | |
| White | |
| Reusable | |
| None | |
| 245 g/m² |
| Cotton | |
| Press Studs | |
| Non-sterile | |
| 2 |