missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lactose octaacetate, 98%

Product Code. 15458149
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458149 5 g 5g
15468149 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458149 Supplier Thermo Scientific Chemicals Supplier No. J66695.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6291-42-5 | C28H38O19 | 678.593 g/mol

Lactose octaacetate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lactose octaacetate is used as pharmaceutical intermediates.

Solubility
Soluble in chloroform

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6291-42-5
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.593
MDL Number MFCD00064040
InChI Key WOTQVEKSRLZRSX-SJSKHYGQSA-N
Synonym lactose octaacetate, 2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-3-2r,3r,4s,5s,6r-3,4,5-tris acetyloxy-6-acetyloxy methyl oxan-2-yl oxy oxan-2-yl methyl acetate, 4-o-2-o,3-o,4-o,6-o-tetraacetyl-alpha-d-galactopyranosyl-beta-d-glucopyranose tetraacetate
PubChem CID 11767585
IUPAC Name [(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-3-[(2R,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in chloroform
Formula Weight 678.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Lactose octaacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.