missing translation for 'onlineSavingsMsg'
Learn More

Lactose Monohydrate, Millipore™

Product Code. 10711882
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10711882 1 kg 1kg
10530761 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10711882 Supplier Millipore Supplier No. 1076601000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10039-26-6

Specifications

Melting Point 202°C
MDL Number MFCD00064074
Quantity 1 kg
Synonym Lactose monohydrate, Milk sugar, Lactobiose, Lactose monohydrate
Molecular Weight (g/mol) 342.30 (anhydrous basis)
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.