missing translation for 'onlineSavingsMsg'
Learn More

Lacidipine, TRC

Product Code. 30135793
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135793 100 mg 100mg
30135549 25 mg 25mg
17941315 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135793 Supplier Toronto Research Chemicals Supplier No. TRCL098000100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103890-78-4
Molecular Formula C26 H33 N O6
Molecular Weight (g/mol) 455.54
Synonym Lacidipine, 4-[2-[(1E)-3-(1,1-Dimethylethoxy)-3-oxo-1-propenyl]phenyl]-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid diethyl ester, (E)-4-[2-[3-(1,1-Dimethylethoxy)-3-oxo-1-propenyl]phenyl]-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid diethyl ester, Caldine, GR 43659X, GX 1048, Lacimen, Lacipil, Lacirex, Midotens, Motens
IUPAC Name diethyl 2,6-dimethyl-4-[2-[(E)-3-[(2-methylpropan-2-yl)oxy]-3-oxoprop-1-enyl]phenyl]-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(C)NC(=C(C1c2ccccc2\C=C\C(=O)OC(C)(C)C)C(=O)OCC)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C26H33NO6/c1-8-31-24(29)21-16(3)27-17(4)22(25(30)32-9-2)23(21)19-13-11-10-12-18(19)14-15-20(28)33-26(5,6)7/h10-15,23,27H,8-9H2,1-7H3/b15-14+
Formula Weight 455.2308
Chemical Name or Material Lacidipine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.