Learn More
Lacidipine, MedChemExpress
Product Code. 16498448
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498448 10 mg 10mg
16498246 100 mg 100mg
16491406 200 mg 200mg
16434337 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16498448 Supplier Medchem Express Supplier No. HYB0347
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
MedChemExpress Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.
Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.
Specifications
Chemical Name or Material Lacidipine
CAS 103890-78-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H33NO6
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (109.76 mM)
SMILES O=C(C1=C(C)NC(C)=C(C(OCC)=O)C1C2=CC=CC=C2/C=C/C(OC(C)(C)C)=O)OCC
Molecular Weight (g/mol) 455.54
Formula Weight 455.54
Percent Purity 99.98%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation