Learn More
Labplas Sani-Sponge Sampling Bags, Regular tabs: With Clear Body, Tear-Off Protection Strip
Product Code. 17976469
Change view
Click to view available options
Includes:
DE Neutralizing buffer
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17976469 DE Neutralizing buffer Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17976469 Supplier Labplas Supplier No. KSS67110DE
  • ONLINE EXCLUSIVE PRICE
    255.00 EUR
    205.53 EUR / Pack of 100
    Save 49.47 EUR
    19% Off
 Restricted Item
Estimated Shipment: 12-03-2027
Designed to collect samples to detect the presence of microbiological contaminations
  • Detects Listeria, Salmonella and E. coli
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
  • Sponge for swabbing
  • Dehydrated sponge
Specifications
Description DE Neutralizing buffer
Includes DE Neutralizing buffer
Volume (Metric) 450 mL
Material Polyethylene