Learn More
Koettermann™ Laboratory Swivel Chair
Product Code. 15397473
Change view
Click to view available options
Height (Metric):
560 to 820 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
15397473 560 to 820 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 15397473 Supplier Koettermann™ Supplier No. 94200002
  • ONLINE EXCLUSIVE PRICE
    514.00 EUR
    283.63 EUR / Pack of 1
    Save 230.37 EUR
    44% Off
Estimated Shipment: 15-01-2027
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Adjustable laboratory swivel chair with runners. Koettermann™ Laboratory Swivel Chair features a PU foam seat and backrest, and a footrest.
  • Adjustable
  • PU foam seat and backrest
  • Footrest
Specifications
Product Type Lab Swivel Chair
Color Black
Adjustable Height
Height (Metric) 560 to 820 mm
Includes Sliders and Foot Support
Footrest Yes