Learn More
Fisherbrand™ Laboratory Paddle Blender with Heat Incubation and UV Chamber Decontamination
Product Code. 17389912 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
400 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17389912 400 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17389912
  • ONLINE EXCLUSIVE PRICE
    6020.00 EUR
    4003.77 EUR / Pack of 1
    Save 2016.23 EUR
    33% Off
Estimated Shipment: 11-12-2026
Fisherbrand Paddle Blender provides fast and effective mixing, ensuring consistent homogenization for food microbiology and other sample prep applications. The chamber can be heated as required. A UV light provides decontamination of the chamber in case of a spill.
  • Contamination free mixing and homogenization
  • Useful for food microbiology and macro sample prep
  • User friendly, touch screen operation
  • Variable speed, time and stroke length
  • Heated chamber for incubated applications
  • Window for visualizing sample
  • Easy to clean
  • UV light for chamber decontamination
  • Accepts 400 mL bags
Specifications
Voltage 220/240 V
Capacity (Metric) 400 mL
Sample Volume (English) 13.5 oz.
No. of Strokes 3 to 12 strokes/sec.
Frequency 60 Hz