Learn More
Epredia™ Laboratory Marking Pen, Secureline Marker II
Product Code. 15582321 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Laboratory Marking Pen
Secureline Marker II
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
15582321 Laboratory Marking Pen Pack of 10
12657736 Secureline Marker II Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15582321 Supplier Epredia™ Supplier No. 2000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Mark cassettes and microscope slides with the Epredia™ Laboratory Marking Pen.
  • Resistant to common laboratory solvents (water, 10% neutral buffered formalin, alcohol)
  • Extra fine point for precise writing
  • Non-sterile
  • Not made with natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Product Type Laboratory Marking Pen
Color Black
Quantity 10/Pk.