Learn More
Fisherbrand™ Laboratory Jacks
Product Code. 11517023 Shop All Fisherbrand Products
Change view
Click to view available options
Height (Metric):
40 to 145 mm (Top Plate)
50 to 270 mm (Top Plate)
60 to 450 mm (Top Plate)
Length (Metric) Base:
110 mm
170 mm
280 mm
Width (Metric) Base:
100 mm
160 mm
260 mm
Capacity (Metric):
15 kg
25 kg
35 kg
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Length (Metric) Base Width (Metric) Base Capacity (Metric) unitSize
11517023 60 to 450 mm (Top Plate) 280 mm 260 mm 35 kg Each
11597013 40 to 145 mm (Top Plate) 110 mm 100 mm 15 kg Each
11507023 50 to 270 mm (Top Plate) 170 mm 160 mm 25 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11517023
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Provides for precise height adjustment with the turn of a knob. Fisherbrand™ Laboratory Jacks are manufactured from aluminum alloy with a lifting screw in stainless steel.
  • Plastic knob operates the scissor action lifting mechanism
  • Either plate can be used as the base
  • Available in a range of plate dimensions
Specifications
Material Aluminum
Height (Metric) 60 to 450 mm (Top Plate)
Length (Metric) Base 280 mm
Width (Metric) Base 260 mm
Capacity (Metric) 35 kg
Includes Stainless steel Lifting screw
Product Type Laboratory Lift
For Use With (Equipment) 12mm dia. Support Rod
Quantity 1 Ea.