Learn More
Labetalol Hydrochloride, TRC
Product Code. 17954385
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17954385 100 mg 100mg
17932875 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17954385 Supplier Toronto Research Chemicals Supplier No. TRCL096495100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32780-64-6
Molecular Formula C19 H24 N2 O3 . Cl H
Molecular Weight (g/mol) 364.87
Synonym Benzamide, 2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, hydrochloride (1:1), Benzamide, 2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, monohydrochloride (9CI), Salicylamide, 5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, monohydrochloride (8CI), AH 5158 hydrochloride, AH 5158A, Amipress, Ipolab, Labelol, Labetalol hydrochloride, Labracol, NSC 290312, Normodyne, Presdate, Pressalolo, Sch 15719W, Trandate, Vescal
IUPAC Name 2-hydroxy-5-[1-hydroxy-2-(4-phenylbutan-2-ylamino)ethyl]benzamide;hydrochloride
SMILES Cl.CC(CCc1ccccc1)NCC(O)c2ccc(O)c(c2)C(=O)N
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C19H24N2O3.ClH/c1-13(7-8-14-5-3-2-4-6-14)21-12-18(23)15-9-10-17(22)16(11-15)19(20)24;/h2-6,9-11,13,18,21-23H,7-8,12H2,1H3,(H2,20,24);1H
Formula Weight 364.1554
Chemical Name or Material Labetalol Hydrochloride