Learn More
Labconco™ Two-Port Manifold for CentriVap™ Systems
Product Code. 15133005 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
CentriVap™ system
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15133005 CentriVap™ system Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15133005 Supplier Labconco™ Supplier No. 7397700
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
Requires
Rotary vane vacuum pump required
Allows freeze-drying of small-volume samples in freeze-dry flasks, serum bottles or ampules
  • Has neoprene rubber valves
  • Connects to CentriVap Cold Trap, Ultra-Low Cold Trap or Mobile System
  • Shipping weight, 1 lb. (0.5kg)
Specifications
Type 2-port Cold Trap Manifold
For Use With (Equipment) CentriVap™ system
Weight (English) 1 lb.
Weight (Metric) 0.5 kg