Learn More
Labconco™ Glassware Cart
Product Code. 16408090 Shop All Labconco Corporation Products
Change view
Click to view available options
No. of Drawers:
2
4
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Drawers Unit Size
16408090 4 Each
15156456 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16408090 Supplier Labconco™ Supplier No. 8045000
  • ONLINE EXCLUSIVE PRICE
    975.00 EUR
    927.68 EUR / Pack of 1
    Save 47.32 EUR
    4% Off
Estimated Shipment: 18-01-2027
Carry glassware, plasticware and small instruments easily with the Labconco Glassware Cart.
Carry glassware, plasticware and small instruments easily with the Labconco Glassware Cart.
  • Load capacity: 400 lb. (181kg)
  • Durable glacier white, powder-coated 18 gauge welded tubular steel frame with ergonomic, extruded rubber handle grips and ID tag
  • Available with two 18L x 32W x 4 in.H (46 x 81 x 10cm) or four 18L x 16W x 4 in.H (46 x 41 x 10cm) removable vinyl-coated wire baskets
  • Removable plastic drip pan catches spills
  • 4 in. dia. (10cm) casters, two with toe locks
  • Lifetime guarantee
Order Info
Shipped fully assembled. Weight: 50 lb. (23kg).
Warranty
Lifetime guarantee.
Specifications
No. of Drawers 4