Learn More
Berkshire Lab-Tips™ Pointed Head Swab
Product Code. 17887290 Shop All Berkshire Products
Change view
Click to view available options
Length (English):
2.75 in.
Unit Size:
Pack of 2000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
17887290 2.75 in. Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17887290 Supplier Berkshire Supplier No. LT070P20
  • ONLINE EXCLUSIVE PRICE
    635.10 EUR
    520.97 EUR / Pack of 2000
    Save 114.13 EUR
    17% Off
View all promotions
Estimated Shipment: 24-02-2027
The LTO70P Swab with pointed head is an excellent swab for precision cleaning applications.
  • Cleanroom laundered polyurethane foam over rigid tip
  • 100 ppi open cell foam
  • Polypropylene handle with thermally bonded head
  • Laundered and packaged in Class 10 {ISO Class 4} cleanroom
  • Low particles, fibers, ions and extractables
  • Good chemical resistance
  • Soft and non-abrasive with no contaminating adhesives
  • Free from Silicone, amides and DOP
  • Ridged surface for improved cleaning ability
Specifications
Length (English) 2.75 in.
Product Type Cleanroom Swab
Autoclavable Not Autoclavable
Adhesive Type Thermal
Handle Material Plastic
Tip Style Pointed
Tip Material Foam