missing translation for 'onlineSavingsMsg'
Learn More

l-Valine, Millipore™

Product Code. 10730022
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10730022 100 g 100g
10172290 25 g 25g
10643471 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10730022 Supplier Millipore Supplier No. 1084950100
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

for biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-18-4

Specifications

Melting Point 315°C
MDL Number MFCD00064220
Synonym L-Valine, Val, L-Valine
Density 1.23 g/cm3 at 20°C
Molecular Weight (g/mol) 117.15
Quantity 100 g
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.