missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine tert-butyl ester, 98%, Thermo Scientific™

Product Code. 15442788
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442788 5 g 5g
15432788 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442788 Supplier Thermo Scientific Alfa Aesar Supplier No. H56322.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16874-12-7
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD00042644
InChI Key DIGHFXIWRPMGSA-LDGXTIHJNA-N
Synonym l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068
PubChem CID 6950582
IUPAC Name tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES CC(C)(C)OC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 141°C to 144°C
Beilstein 2696011
Quantity 5 g
Formula Weight 237.29
Percent Purity 98%
Chemical Name or Material L-Tyrosine tert-butyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.