Learn More
L-Tryptophanamide Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | L-Tryptophanamide Hydrochloride |
| CAS | 5022-65-1 |
| Molecular Formula | C11 H13 N3 O . Cl H |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C11H13N3O.ClH/c12-9(11(13)15)5-7-6-14-10-4-2-1-3-8(7)10;/h1-4,6,9,14H,5,12H2,(H2,13,15);1H/t9-;/m0./s1 |
| SMILES | Cl.N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N |
| IUPAC Name | (2S)-2-amino-3-(1H-indol-3-yl)propanamide;hydrochloride |
| Molecular Weight (g/mol) | 239.7 |
| Formula Weight | 239.0825 |