missing translation for 'onlineSavingsMsg'
Learn More

l-Tryptophan, Millipore™

Product Code. 10683461
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10683461 500 g 500g
10553861 10 g 10g
10077310 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10683461 Supplier Millipore Supplier No. 1083740500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-22-3

Specifications

Melting Point 290°C (decomposition)
MDL Number MFCD00064340
Synonym L-Tryptophan, Trp, L-Tryptophan
Molecular Weight (g/mol) 204.23
Quantity 500 g
Grade Research
Chemical Name or Material L-Tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.