missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan Ethyl Ester Hydrochloride, Mikromol™

Product Code. 30462082
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462082 4 x 25 mg -
30049088 25 mg 25mg
30051904 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30462082 Supplier Mikromol Supplier No. MM0172.1800254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Tryptophan Ethyl Ester Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Tryptophan Ethyl Ester Hydrochloride
CAS 2899-28-7
Analyte or Component Names L-Tryptophan Ethyl Ester Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H16 N2 O2 . Cl H
Certifications/Compliance ISO 9001
Synonym L-Tryptophan Ethyl Ester Hydrochloride, L-Tryptophan, ethyl ester hydrochloride (1:1), L-Tryptophan, ethyl ester, monohydrochloride (9CI), Tryptophan, ethyl ester, monohydrochloride, L- (8CI), (S)-Tryptophan ethyl ester hydrochloride, H-Trp-OEt•HCl, L-Tryptophan ethyl ester hydrochloride
InChI Formula InChI=1S/C13H16N2O2.ClH/c1-2-17-13(16)11(14)7-9-8-15-12-6-4-3-5-10(9)12;/h3-6,8,11,15H,2,7,14H2,1H3;1H/t11-;/m0./s1
Solution Type Neat
SMILES Cl.CCOC(=O)[C@@H](N)Cc1c[nH]c2ccccc12
IUPAC Name ethyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
Molecular Weight (g/mol) 268.74
Quantity 4 x 25 mg
Formula Weight 268.0979 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.