missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tryptophan Ethyl Ester Hydrochloride, Mikromol™
Description
L-Tryptophan Ethyl Ester Hydrochloride
Specifications
Specifications
| Chemical Name or Material | L-Tryptophan Ethyl Ester Hydrochloride |
| CAS | 2899-28-7 |
| Analyte or Component Names | L-Tryptophan Ethyl Ester Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H16 N2 O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Tryptophan Ethyl Ester Hydrochloride, L-Tryptophan, ethyl ester hydrochloride (1:1), L-Tryptophan, ethyl ester, monohydrochloride (9CI), Tryptophan, ethyl ester, monohydrochloride, L- (8CI), (S)-Tryptophan ethyl ester hydrochloride, H-Trp-OEt•HCl, L-Tryptophan ethyl ester hydrochloride |
| InChI Formula | InChI=1S/C13H16N2O2.ClH/c1-2-17-13(16)11(14)7-9-8-15-12-6-4-3-5-10(9)12;/h3-6,8,11,15H,2,7,14H2,1H3;1H/t11-;/m0./s1 |
| SMILES | Cl.CCOC(=O)[C@@H](N)Cc1c[nH]c2ccccc12 |
| IUPAC Name | ethyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?