Learn More
L-Tryptophan-d5, TRC
Product Code. 30456730
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456730 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30456730 Supplier Toronto Research Chemicals Supplier No. TRCT947212100MG
 Restricted Item
Estimated Shipment: 11-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material L-Tryptophan-d5
CAS 62595-11-3
Purity Grade Notes HPLC
Molecular Formula C11 2H5 H7 N2 O2
Quantity 100 mg
Synonym L-Tryptophan-2,4,5,6,7-D5, L-(2,4,5,6,7-2H5)Tryptophan, L-Tryptophan-d5, (S)-Tryptophan-d5, (S)-α-Amino-3-(indole-2,4,5,6,7-d5)propionic acid, (S)-α-Amino(indole-2,4,5,6,7-d5)-3-propanoic acid
InChI Formula InChI=1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1/i1D,2D,3D,4D,6D
SMILES [2H]c1[nH]c2c([2H])c([2H])c([2H])c([2H])c2c1C[C@H](N)C(=O)O
IUPAC Name (2S)-2-amino-3-(2,4,5,6,7-pentadeuterio-1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 209.26
Formula Weight 209.1213
Percent Purity >95