missing translation for 'onlineSavingsMsg'
Learn More

L-Thryoxine, 98.5%, MP Biomedicals™

Product Code. 11468301 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.1 g
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468301 0.1 g 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11468301 Supplier MP Biomedicals Supplier No. 02152145.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 51-48-9
Color White to Yellow
Assay Percent Range 0.985
Molecular Formula C15H11I4NO4
MDL Number MFCD00002595
Quantity 0.1 g
Synonym l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum
InChI Key XUIIKFGFIJCVMT-UHFFFAOYNA-N
SMILES NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O
IUPAC Name 2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
Molecular Weight (g/mol) 776.87
PubChem CID 5819
ChEBI CHEBI:18332
Formula Weight 776.9
Physical Form Crystalline Powder
Chemical Name or Material L-Thryoxine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.