Learn More
L-Threonine, Millipore™
Product Code. 32238857
Change view
Click to view available options
Quantity:
10 kg
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238857 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238857 Supplier Millipore Supplier No. 1084119010
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
For biochemistry
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material L-Threonine
Melting Point 265°C to 270°C
CAS 72-19-5
Synonym L-Threonine, Thr, 2-Amino-3-hydroxybutanoic acid, L-Threonine
Molecular Weight (g/mol) 119.12
Quantity 10 kg