missing translation for 'onlineSavingsMsg'
Learn More

l-Proline, Millipore™

Product Code. 10712262
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
10 kg
Unit Size:
100g
10g
10kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10712262 10 kg 10kg
10539050 10 g 10g
10153550 100 g 100g
10177600 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10712262 Supplier Millipore Supplier No. 1074349010
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

for biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-85-3

Specifications

Melting Point 220°C to 222°C (decomposition)
MDL Number MFCD00064318
Synonym L-Proline, Pro, L-Proline
Density 1.35 to 1.38 g/cm3 at 25°C
Molecular Weight (g/mol) 115.13
Quantity 10 kg
Grade Research
Chemical Name or Material L-Proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.