missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine, >99%, MP Biomedicals

Product Code. 11411981 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
missing translation for 'unitSize'
100g
1kg
250g
25g
500g
This item is not returnable. View return policy

Product Code. 11411981

missing translation for 'mfr': MP Biomedicals 0219472391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Quantity 1 kg
Formula Weight 165.2
Grade Cell Culture
Chemical Name or Material L-Phenylalanine, >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.